Tetrabutylammonium tetrafluoroborate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetrabutylammoniumtetrafluoroborate

A16688-06 A16688-14
CAAAA16688-06EA 65.73 CAD
CAAAA16688-06 CAAAA16688-14
Tetrabutylammonium tetrafluoroborate 99%
Tetrabutylammonium tetrafluoroborate
Formula: C₁₆H₃₆BF₄N
MW: 329.27 g/mol
Melting Pt: 159…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00011634
CAS Number: 429-42-5
EINECS: 207-058-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR