Lanthanum(III) fluoride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lanthanum trifluoride

A16673-14 A16673-22
CAAAAA16673-14EA 74.73 CAD
CAAAAA16673-14 CAAAAA16673-22
Lanthanum(III) fluoride 99%
Lanthanum (III) fluoride

Purity: 99% CAS No.: 13709-38-1 Molecular Formula: F3La Formula Weight: 195.90 Odor: Odorless Melting Point: 1493° MDL No.: MFCD00011073

Formula: F₃La
MW: 195.9 g/mol
Boiling Pt: ~ 2327 °C (1013 hPa)
Melting Pt: 1493 °C
Density: 5.936
Storage Temperature: Ambient
MDL Number: MFCD00011073
CAS Number: 13709-38-1
EINECS: 237-252-8
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR