Potassium carbonate, anhydrous 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Potash

A16625-0C A16625-0E A16625-36
CAAAAA16625-0CEA 440.82 CAD
CAAAAA16625-0C CAAAAA16625-0E CAAAAA16625-36
Potassium carbonate, anhydrous 99%
Potassium carbonate

Purity: 99% CAS No.: 584-08-7 Molecular Formula: CK2O3 Formula Weight: 138.21 Odor: Odorless pH: 11,5-12,5 Melting Point: 891° Beilstein Reference: 4267587 Merck Reference: 14,7619 MDL No.: MFCD00011382

Formula: K₂CO₃
Melting Pt: 891 °C
Density: 2.29
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 14,7619
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR