Orthoboric acid 98%

Supplier: Thermo Scientific Chemicals
Danger

A16624-0B A16624-0I
CAAAAA16624-0BEA 79.32 CAD
CAAAAA16624-0B CAAAAA16624-0I
Orthoboric acid 98%
ortho-Boric acid

Purity: 98% CAS No.: 10043-35-3 Molecular Formula: BH3O3 Formula Weight: 61.83 Flash Point: None Odor: Odorless pH: 3.8-4.8 Melting Point: ca 185° dec. Beilstein Reference: 1697939 Merck Reference: 14,1336 MDL No.: MFCD00011337

Formula: B(OH)₃
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: ∼ 185 °C (dec.)
Density: 1.435
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 14,1336
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR