2-Methyl-2-pentenoic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

A16587-09 A16587-18 A16587-30
CAAAA16587-09EA 52.46 CAD
CAAAA16587-09 CAAAA16587-18 CAAAA16587-30
2-Methyl-2-pentenoic acid 99%
2-Methyl-2-pentenoic acid

Purity: 99% CAS No.: 3142-72-1 Molecular Formula: C6H10O2 Formula Weight: 114.15 Flash Point: 107°(225°F) Odor: Characteristic Boiling Point: 120°/20mm UN No.: 3265 MDL No.: MFCD00002655

Formula: C₆H₁₀O₂
Boiling Pt: 120 °C/20 mm
Density: 0.978
Flash Pt: 107 °C(225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002655
CAS Number: 3142-72-1
EINECS: 221-552-0
UN: 3265
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR