1-Chloro-3-methyl-2-butene 95% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with potassium carbonate
A16545-09
A16545-18
CAAAA16545-09EA
130.67
CAD
CAAAA16545-09
CAAAA16545-18
1-Chloro-3-methyl-2-butene 95% stabilized
1-Chloro-3-methyl-2-butene
Purity: 95%
CAS No.: 503-60-6
Molecular Formula: C5H9Cl
Formula Weight: 104.57
Storage Temperature: 2°C to 8°C
Flash Point: 13°(56°F)
Refractive Index: 1.4490
Boiling Point: 107-109°
UN No.: 1993
MDL No.: MFCD00000982
Formula:
C₅H₉Cl MW: 104.58 g/mol Boiling Pt: 107…109 °C Density: 0.928 Flash Pt: 13 °C (56 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000982 CAS Number: 503-60-6 EINECS: 207-972-7 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...