2,3-Dimethylphenylhydrazinium hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16526-06
A16526-14
CAAAAA16526-36EA
956.36
CAD
CAAAAA16526-36
CAAAA16526-06
CAAAA16526-14
2,3-Dimethylphenylhydrazinium hydrochloride 97%
2,3-Dimethylphenylhydrazinium hydrochloride
Purity: 97%
CAS No.: 56737-75-8
Molecular Formula: C8H12N2.HCl
Formula Weight: 172.66
Melting Point: ca 210° dec.
Beilstein Reference: 6096287
Formula:
C₈H₁₃ClN₂ Melting Pt: ca 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013380 CAS Number: 56737-75-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...