Dibenzofuran 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dibenzo[b,d]furan
A16521-14
A16521-22
CAAAAA16521-14EA
144.57
CAD
CAAAAA16521-14
CAAAAA16521-22
Dibenzofuran 98%
Dibenzofuran
Purity: 98%
CAS No.: 132-64-9
Molecular Formula: C12H8O
Formula Weight: 168.20
Flash Point: 130°(266°F)
Melting Point: 78-83°
Boiling Point: 287-289°
Beilstein Reference: 121100
UN No.: 3077
MDL No.: MFCD00004968
Formula:
C₁₂H₈O MW: 168.19 g/mol Boiling Pt: 287…289 °C Melting Pt: 78…83 °C Density: 1.3 Flash Pt: 130 °C (266 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004968 CAS Number: 132-64-9 EINECS: 205-071-3 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...