Cobalt(II) chloride hexahydrate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16346-0B
A16346-18
A16346-30
CAAAAA16346-0BEA
418.7
CAD
CAAAAA16346-0B
CAAAAA16346-18
CAAAAA16346-30
Cobalt(II) chloride hexahydrate 98%
Cobalt (II) chloride hexahydrate
Purity: 98%
CAS No.: 7791-13-1
Molecular Formula: Cl2Co.6H2O
Formula Weight: 237.93 (129.84anhy)
Odor: Odorless
pH: 4.9
Melting Point: 87°
Merck Reference: 14,2437
UN No.: 3077
MDL No.: MFCD00149652
Formula:
CoCl₂·6H₂O Boiling Pt: 110 °C (1013 hPa) Melting Pt: 87 °C Density: 1.924 Storage Temperature: Ambient |
MDL Number:
MFCD00149652 CAS Number: 7791-13-1 EINECS: 231-589-4 UN: 3077 ADR: 9,III Merck Index: 14,2437 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...