Lead(II) nitrate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead dinitrate

A16345-0E A16345-22 A16345-36
CAAAAA16345-0EEA 363.4 CAD
CAAAAA16345-0E CAAAAA16345-22 CAAAAA16345-36
Lead(II) nitrate 99%
Lead (II) nitrate

Purity: 99% CAS No.: 10099-74-8 Molecular Formula: Pb(NO3)2 Formula Weight: 331.20 Odor: Odorless Melting Point: 470° UN No.: 1469 MDL No.: MFCD00011153

Formula: Pb(NO₃)₂
MW: 331.21 g/mol
Melting Pt: 470 °C
Density: 4.530
Storage Temperature: Ambient
MDL Number: MFCD00011153
CAS Number: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1 II
Merck Index: 14,5414

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR