Ammonium acetate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetic acid ammonium salt

A16343-0B A16343-0I A16343-30
CAAAAA16343-0BEA 128.93 CAD
CAAAAA16343-0B CAAAAA16343-0I CAAAAA16343-30
Ammonium acetate 97%
Ammonium acetate

Purity: 97% CAS No.: 631-61-8 Molecular Formula: C2H7NO2 Formula Weight: 77.08 Flash Point: 136°(277°F) Odor: Ammonia-like Boiling Point: dec. Beilstein Reference: 4186741 Merck Reference: 14,495 MDL No.: MFCD00013066

Formula: H₃CCOONH₄
MW: 77.08 g/mol
Boiling Pt: Dec.
Melting Pt: 114 (dec.)
Density: 1.17
Flash Pt: 136 °C (277 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 14,495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form Crystals or powder or crystalline powder
Identification (FTIR) Conforms
Ammonia as ammonium acetate ≥96.0% to ≤104.0%
Water Content (Karl Fischer Titration) ≤5.0%
Appearance (Color) White


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR