Potassium bromide 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16339-0B
A16339-0I
A16339-22
A16339-36
A16339-A4
CAAAAA16339-0BEA
191.18
CAD
CAAAAA16339-0B
CAAAAA16339-0I
CAAAAA16339-22
CAAAAA16339-36
CAAAAA16339-A4
Potassium bromide 99+%
Potassium bromide
Purity: 99+%
CAS No.: 7758-02-3
Molecular Formula: KBr
Formula Weight: 119.01
Refractive Index: 1.559
Odor: Odorless
Boiling Point: 1435°
Merck Reference: 14,7618
MDL No.: MFCD00011358
Formula:
KBr Boiling Pt: 1435 °C Melting Pt: 730 °C Density: 2.75 Storage Temperature: Ambient |
MDL Number:
MFCD00011358 CAS Number: 7758-02-3 EINECS: 231-830-3 Merck Index: 14,7618 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...