Iron(II) chloride tetrahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ferrous chloride tetrahydrate, Iron dichloride tetrahydrate

A16327-0B A16327-30
CAAAAA16327-0BEA 320.74 CAD
CAAAAA16327-0B CAAAAA16327-30
Iron(II) chloride tetrahydrate 98%
Iron (II) chloride tetrahydrate

Purity: 98% CAS No.: 13478-10-9 Molecular Formula: Cl2Fe.4H2O Formula Weight: 198.81 (126.75anhy) Odor: Odorless Merck Reference: 14,4043 MDL No.: MFCD00149709

Formula: FeCl₂·4H₂O
Melting Pt: 677 °C
Density: 1.93
Storage Temperature: Ambient
MDL Number: MFCD00149709
CAS Number: 13478-10-9
EINECS: 231-843-4
UN: 3260
ADR: 8,III
Merck Index: 14,4043

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR