Potassium acetate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Acetic acid potassium salt

A16321-0C A16321-0E A16321-36
CAAAAA16321-0CEA 917.98 CAD
CAAAAA16321-0C CAAAAA16321-0E CAAAAA16321-36
Potassium acetate 99%
Potassium acetate

Purity: 99% CAS No.: 127-08-2 Molecular Formula: CH3CO2K Formula Weight: 98.15 Melting Point: 292° Beilstein Reference: 3595449 Merck Reference: 14,7605 MDL No.: MFCD00012458

Formula: CH₃COOK
Melting Pt: 292 °C
Density: 1.57
Flash Pt: ≥ 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 14,7605
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR