Sodium decyl sulfate 99%

Supplier: Thermo Scientific Chemicals
Danger

A16299-03 A16299-06 A16299-14
CAAAAA16299-03EA 97.8 CAD
CAAAAA16299-03 CAAAAA16299-06 CAAAAA16299-14
Sodium decyl sulfate 99%
Sodium decyl sulphate

Purity: 99% CAS No.: 142-87-0 Molecular Formula: CH3(CH2)9OSO3Na Formula Weight: 260.33 Beilstein Reference: 3576860 MDL No.: MFCD00041881

Formula: C₁₀H₂₁NaO₄S
Melting Pt: 191,85 °C
Density: 1,207 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041881
CAS Number: 142-87-0
EINECS: 205-568-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR