Zinc sulfide 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Zinc (II) sulphide, Zinc (II) sulfide

A16296-0B A16296-0I A16296-30
CAAAAA16296-0BEA 188.02 CAD
CAAAAA16296-0B CAAAAA16296-0I CAAAAA16296-30
Zinc sulfide 98%
Zinc sulphide

Purity: 98% CAS No.: 1314-98-3 Molecular Formula: SZn Formula Weight: 97.43 Odor: Odorless Melting Point: 1700° MDL No.: MFCD00011301

Formula: SZn
MW: 97.46 g/mol
Melting Pt: 1700 °C
Density: 4.10
Storage Temperature: Ambient
MDL Number: MFCD00011301
CAS Number: 1314-98-3
EINECS: 215-251-3
Merck Index: 14,10160

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR