Zinc chloride, anhydrous 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride

A16281-0C A16281-0E A16281-22 A16281-36
CAAAAA16281-0CEA 1388.82 CAD
CAAAAA16281-0C CAAAAA16281-0E CAAAAA16281-22 CAAAAA16281-36
Zinc chloride, anhydrous 98+%
Zinc chloride

Purity: 98+% CAS No.: 7646-85-7 Molecular Formula: Cl2Zn Formula Weight: 136.29 Odor: Odorless Melting Point: 290° Boiling Point: 732° Merck Reference: 14,10132 UN No.: 2331 MDL No.: MFCD00011295

Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732
Melting Pt: 283
Density: 2.91
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8 III
Merck Index: 14,10132

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR