Calcium hydride ∼92%, coarse powder
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16242-0B
A16242-18
A16242-30
CAAAAA16242-0BEA
1087.04
CAD
CAAAAA16242-0B
CAAAAA16242-18
CAAAAA16242-30
Calcium hydride ∼92%, coarse powder
Calcium hydride
Purity: 92%
CAS No.: 7789-78-8
Molecular Formula: CaH2
Formula Weight: 42.10
Odor: Odorless
Melting Point: 600° dec.
UN No.: 1404
MDL No.: MFCD00010897
Formula:
H₂Ca Melting Pt: 600 °C (dec.) Density: 1.900 Storage Temperature: Ambient |
MDL Number:
MFCD00010897 CAS Number: 7789-78-8 EINECS: 232-189-2 UN: 1404 ADR: 4.3,I Merck Index: 14,1672 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...