4-(Methylamino)phenol sulfate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-(Methylamino)phenol sulphate, Metol, 4-(Methylamino)phenol hemisulfate, 4-(Methylamino)phenol hemisulfate salt, p-(Methylamino)phenol sulfate, 4-Methylaminophenol sulphate, 4-Methylaminophenolsulfate, 4-(Methylamino)phenol hemisulphate, 4-Methylaminophenol sulfate, 4-Methylaminophenolsulphate, 4-(Methylamino)phenol hemisulphate salt

A16236-0B A16236-18 A16236-30
CAAAAA16236-0BEA 423.44 CAD
CAAAAA16236-0B CAAAAA16236-18 CAAAAA16236-30
4-(Methylamino)phenol sulfate 99%
4-(Methylamino)phenol sulfate

Purity: 99% CAS No.: 55-55-0 Molecular Formula: C14H18N2O2.H2SO4 Formula Weight: 344.38 Melting Point: ca 260° dec. Beilstein Reference: 3919382 Merck Reference: 14,6017 UN No.: 2811 MDL No.: MFCD00013140

Formula: C₁₄H₂₀N₂O₆S
MW: 344.39 g/mol
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: ∼ 260 °C (dec.)
Density: 1.434 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013140
CAS Number: 55-55-0
EINECS: 200-237-1
UN: 2811
ADR: 6.1,III
Merck Index: 14,6017

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR