Iron(III) chloride hexahydrate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Iron trichloride hexahydrate, Ferric chloride hexahydrate

A16231-0C A16231-0E A16231-36
CAAAAA16231-0CEA 1360.38 CAD
CAAAAA16231-0C CAAAAA16231-0E CAAAAA16231-36
Iron(III) chloride hexahydrate 97%
Iron (III) chloride hexahydrate

Purity: 97% CAS No.: 10025-77-1 Molecular Formula: Cl3Fe.6H2O Formula Weight: 270.30 (162.21anhy) Odor: Odorless pH: 1.8 Melting Point: 37° Boiling Point: 280-285° Merck Reference: 14,4019 UN No.: 3260 MDL No.: MFCD00149712

Formula: FeCl₃·6H₂O
MW: 270.3 g/mol
Boiling Pt: 280…285 °C
Melting Pt: 37 °C
Density: 1.820
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1
EINECS: 231-729-4
UN: 3260
ADR: 8,III
Merck Index: 14,4019
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=808c75376e55f29296d58eb1e5a1e184039ce733&vid=6d820873011bee3ff401441544f7f85599e40626&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR