Sodium acetate trihydrate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Acetic acid sodium salt trihydrate
A16230-0C
A16230-0E
A16230-36
CAAAAA16230-0CEA
810.54
CAD
CAAAAA16230-0C
CAAAAA16230-0E
CAAAAA16230-36
Sodium acetate trihydrate 99%
Sodium acetate trihydrate
Purity: 99%
CAS No.: 6131-90-4
Molecular Formula: NaOOCCH3.3H2O
Formula Weight: 136.08 (82.03anhy)
Odor: Odorless
pH: 7.5-9.0
Melting Point: 62-64°
Beilstein Reference: 3732037
Merck Reference: 14,8571
MDL No.: MFCD00071557
Formula:
H₃CCOONa·3H₂O Boiling Pt: 120 - 123 °C (1013 hPa) Melting Pt: 62…64 °C Density: 1.45 Storage Temperature: Ambient |
MDL Number:
MFCD00071557 CAS Number: 6131-90-4 EINECS: 204-823-8 Merck Index: 14,8571 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...