Copper(II) acetate monohydrate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16203-0B
A16203-0I
A16203-30
CAAAAA16203-0BEA
225.94
CAD
CAAAAA16203-0B
CAAAAA16203-0I
CAAAAA16203-30
Copper(II) acetate monohydrate 98+%
Copper (II) acetate monohydrate
Purity: 98+%
CAS No.: 6046-93-1
Molecular Formula: C4H6CuO4.H2O
Formula Weight: 199.65 (181.64anhy)
Physical Form: Powder
Density: 1.882
Appearance: Blue-green
Odor: Odorless
pH: 5.2-5.5
Melting Point: 115°
Merck Reference: 14,2624
UN No.: 3077
MDL No.: MFCD00149570
Formula:
(H₃CCOO)₂Cu·H₂O Melting Pt: 115 °C Density: 1.882 Storage Temperature: Ambient |
MDL Number:
MFCD00149570 CAS Number: 6046-93-1 EINECS: 205-553-3 UN: 3260 ADR: 8,III Merck Index: 14,2624 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...