Cobalt(II) sulfate heptahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cobalt sulphate heptahydrate, Cobaltous sulphate heptahydrate, Cobalt sulfate heptahydrate, Cobaltous sulfate heptahydrate

A16201-0E A16201-22 A16201-36
CAAAAA16201-0EEA 635.16 CAD
CAAAAA16201-0E CAAAAA16201-22 CAAAAA16201-36
Cobalt(II) sulfate heptahydrate 98%
Cobalt (II) sulphate heptahydrate

Purity: 98% CAS No.: 10026-24-1 Molecular Formula: CoO4S.7H2O Formula Weight: 281.10 (154.99anhy) Odor: Odorless pH: 4 Merck Reference: 14,2448 UN No.: 3077 MDL No.: MFCD00149658

Formula: CoSO₄·7H₂O
MW: 281.1 g/mol
Melting Pt: 735
Density: 2.03
Storage Temperature: Ambient
MDL Number: MFCD00149658
CAS Number: 10026-24-1
EINECS: 233-334-2
UN: 3077
ADR: 9 III
Merck Index: 14,2448

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR