Potassium hydroxide 85, flake

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potash caustic, Caustic potash

A16199-36 A16199-0C A16199-0E
CAAAAA16199-36EA 56.94 CAD
CAAAAA16199-36 AAA16199-0C AAA16199-0E
Potassium hydroxide 85, flake
Potassium hydroxide

Purity: 85% CAS No.: 1310-58-3 Molecular Formula: HKO Formula Weight: 56.11 Odor: Odorless pH: 13,5 Melting Point: 360° Boiling Point: 1320-1324° Merck Reference: 14,7640 UN No.: 1813 MDL No.: MFCD00003553

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 2.044
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
REACH: 01-2119487136-33
Merck Index: 14,7640

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR