Barium diphenylamine-4-sulfonate

Supplier: Thermo Scientific Chemicals
Warning

A16197-06 A16197-14
CAAAAA16197-06EA 111.55 CAD
CAAAAA16197-06 CAAAAA16197-14
Barium diphenylamine-4-sulfonate
Barium diphenylamine-4-sulphonate

CAS No.: 6211-24-1 Molecular Formula: C24H20BaN2O6S2 Formula Weight: 633.90 Melting Point: 300° UN No.: 1564 MDL No.: MFCD00007497

Formula: C₂₄H₂₀BaN₂O₆S₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007497
CAS Number: 6211-24-1
EINECS: 228-278-0
UN: 1564
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c37e55aa4075d4ea7537fbef23f9195faa03f14&vid=5a934374d2d4e697c6ecd2cd9828f06692feca01&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR