Tetrasodium diphosphate decahydrate 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium pyrophosphate decahydrate, tetra-Sodium pyrophosphate decahydrate, Sodium pyrophosphate tetrabasic decahydrate, TSPP
A16195-0B
A16195-0I
A16195-30
CAAAAA16195-0BEA
141.88
CAD
CAAAAA16195-0B
CAAAAA16195-0I
CAAAAA16195-30
Tetrasodium diphosphate decahydrate 97+%
tetra-Sodium diphosphate decahydrate
Purity: 97+%
CAS No.: 13472-36-1
Molecular Formula: Na4O7P2.10H2O
Formula Weight: 446.06 (265.90anhy)
Odor: Odorless
pH: 9.5-10.5
Melting Point: 79.5°
Merck Reference: 14,9240
MDL No.: MFCD00149200
Formula:
Na₄P₂O₇·10H₂O MW: 446.06 g/mol Melting Pt: 990 Density: 1.82 Storage Temperature: Ambient |
MDL Number:
MFCD00149200 CAS Number: 13472-36-1 EINECS: 231-767-1 Merck Index: 14,9240 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...