di-Sodium L(+)-tartrate dihydrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium L-(+)-tartrate dihydrate, L(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Sodium tartrate dibasic dihydrate, Disodium L-(+)-Tartrate Dihydrate, (R,R)-(+)-Disodium tartrate dihydrate, L(+)-Tartaric acid, disodium salt, dihydrate, Sodium L-tartrate dibasic dihydrate, L(+)-Sodium tartrate dibasic dihydrate, Sodium Tartrate, Dihydrate, (+)-Disodium L-tartrate dihydrate, (+)-Disodium-L-tartrat dihydrate

A16187-0B A16187-0I A16187-30
CAAAAA16187-0BEA 241.74 CAD
CAAAAA16187-0B CAAAAA16187-0I CAAAAA16187-30
di-Sodium L(+)-tartrate dihydrate 99%
di-Sodium L(+)-tartrate dihydrate

Purity: 99% CAS No.: 6106-24-7 Molecular Formula: C4H4Na2O6.2H2O Formula Weight: 230.08 (194.06anhy) Odor: Odorless Melting Point: ca 150° -2H2O Beilstein Reference: 6121732 Merck Reference: 14,8684 MDL No.: MFCD00150035

Formula: NaOOC(CHOH)₂COONa·2H₂O
MW: 230.08 g/mol
Melting Pt: 57 (–H₂O of crystall.)
Density: 1.82
Storage Temperature: Ambient
MDL Number: MFCD00150035
CAS Number: 6106-24-7
EINECS: 212-773-3
Merck Index: 14,8684
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39a75038f4ff5e6c234267b15540823ae361bed4&vid=89289572c48b0c6415f8f11e916a51f66e129474&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR