di-Sodium tetraborate decahydrate 99+%

Supplier: Thermo Scientific Chemicals
Danger

A16176-0C A16176-0E A16176-36
CAAAAA16176-0CEA 485.06 CAD
CAAAAA16176-0C CAAAAA16176-0E CAAAAA16176-36
di-Sodium tetraborate decahydrate 99+%
di-Sodium tetraborate decahydrate

Purity: 99+% CAS No.: 1303-96-4 Molecular Formula: B4Na2O7.10H2O Formula Weight: 381.37 (201.22anhy) Odor: Odorless pH: 9,24 Melting Point: 75° Merck Reference: 14,8590 MDL No.: MFCD00149193

Formula: Na₂B₄O₇·10H₂O
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 75 °C
Density: 1.73
Storage Temperature: Ambient
MDL Number: MFCD00149193
CAS Number: 1303-96-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 14,8590

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR