2-Furaldehyde 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Furfurol, Furfural

A16167-0B A16167-30
CAAAAA16167-0BEA 94.8 CAD
CAAAAA16167-0B CAAAAA16167-0I CAAAAA16167-30
2-Furaldehyde 98%
2-Furaldehyde

Purity: 98% CAS No.: 98-01-1 Molecular Formula: C5H4O2 Formula Weight: 96.09 Flash Point: 60°(140°F) Refractive Index: 1.5261 Odor: Almond-like Melting Point: -38° Boiling Point: 162° Beilstein Reference: 105755 Merck Reference: 14,4304 UN No.: 1199 MDL No.: MFCD00003229

Formula: C₅H₄O₂
MW: 96.09 g/mol
Boiling Pt: 162 °C
Melting Pt: –38 °C
Density: 1.159
Flash Pt: 60 °C (140 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003229
CAS Number: 98-01-1
EINECS: 202-627-7
UN: 1199
ADR: 6.1,II
Merck Index: 14,4304

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR