Formaldehyde 37% w/w aqueous solution Stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
Stabilization:
7-8% MeOH
A16163-0F
A16163-AP
CAAAAA16163-0FEA
109.08
CAD
CAAAAA16163-0F
CAAAAA16163-AE
CAAAAA16163-AP
Formaldehyde 37% w/w aqueous solution Stabilised
Formaldehyde
CAS No.: 50-00-0
Molecular Formula: CH2O
Formula Weight: 30.03
Flash Point: 60°(140°F)
Refractive Index: 1.3765
Odor: Pungent
Boiling Point: 97°
Beilstein Reference: 1209228
Merck Reference: 14,4235
UN No.: 1198
MDL No.: MFCD00003274
Formula:
CH₂O MW: 30.03 g/mol Boiling Pt: -19.3 Melting Pt: -92 Density: 1.083 Flash Pt: 66 °C (151 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 EINECS: 200-001-8 UN: 1198 ADR: 3 III Merck Index: 14,4235 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...