Iodine monobromide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16158-14
A16158-22
A16158-30
CAAAAA16158-14EA
74.73
CAD
CAAAAA16158-14
CAAAAA16158-22
CAAAAA16158-30
Iodine monobromide 98%
Iodine monobromide
Purity: 98%
CAS No.: 7789-33-5
Molecular Formula: IBr
Formula Weight: 206.81
Storage Temperature: 2°C to 8°C
Flash Point: None
Odor: Pungent
Melting Point: 50° subl.
Boiling Point: 116° dec.
Beilstein Reference: 3902973
Merck Reference: 14,5016
UN No.: 3260
MDL No.: MFCD00011353
Formula:
IBr Boiling Pt: 116 °C (dec.) Melting Pt: 50 °C (subl.) Density: 4.416 Storage Temperature: Refrigerator |
MDL Number:
MFCD00011353 CAS Number: 7789-33-5 EINECS: 232-159-9 UN: 3260 ADR: 8,III Merck Index: 14,5016 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...