Ammonium iron (III) sulfate dodecahydrate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16105-0E
A16105-36
CAAAAA16105-0EEA
295.46
CAD
CAAAAA16105-0E
CAAAAA16105-36
Ammonium iron (III) sulfate dodecahydrate 98+%
Ammonium iron (III) sulphate dodecahydrate
Purity: 98+%
CAS No.: 7783-83-7
Molecular Formula: H4FeNO8S2.12H2O
Formula Weight: 482.18 (266.00anhy)
Odor: Odorless
Melting Point: 39-41°
MDL No.: MFCD00150004
Formula:
NH₄Fe(SO₄)₂·12H₂O Boiling Pt: 230 °C (1013 hPa) Melting Pt: 39…41 °C Density: 1.71 Storage Temperature: Ambient |
MDL Number:
MFCD00150004 CAS Number: 7783-83-7 EINECS: 233-382-4 Merck Index: 14,521 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...