Lithium dodecyl sulfate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dodecyl sulphuric acid lithium salt, Dodecyl sulfate lithium salt, Dodecyl sulphate lithium salt, Lithium lauryl sulphate, Lauryl sulfate lithium salt, Dodecyl sulfuric acid lithium salt, Lithium lauryl sulfate, Lauryl sulphate lithium salt
A16005-06
A16005-14
A16005-22
CAAAAA16005-06EA
66.68
CAD
CAAAAA16005-06
CAAAAA16005-14
CAAAAA16005-22
Lithium dodecyl sulfate 98%
Lithium dodecyl sulphate
Purity: 98%
CAS No.: 2044-56-6
Molecular Formula: CH3(CH2)11OSO3Li
Formula Weight: 272.33
Odor: Odorless
UN No.: 1325
MDL No.: MFCD00007467
Formula:
C₁₂H₂₅LiO₄S MW: 272.34 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007467 CAS Number: 2044-56-6 EINECS: 218-058-2 UN: 1325 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...