trans-2-Octen-1-ol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15960-06
A15960-14
A15960-22
CAAAAA15960-06EA
66.52
CAD
CAAAAA15960-06
CAAAAA15960-14
CAAAAA15960-22
trans-2-Octen-1-ol 97%
trans-2-Octen-1-ol
Purity: 97%
CAS No.: 18409-17-1
Molecular Formula: C8H16O
Formula Weight: 128.22
Flash Point: 85°(185°F)
Refractive Index: 1.4465
Boiling Point: 88°/11mm
Beilstein Reference: 1720707
MDL No.: MFCD00014057
Formula:
C₈H₁₆O Boiling Pt: 88 °C/11 mm Density: 0.850 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014057 CAS Number: 18409-17-1 EINECS: 242-288-2 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...