Thiomorpholine 98%

Supplier: Thermo Scientific Chemicals
Danger

A15958-06 A15958-14 A15958-22
CAAAA15958-06EA 100.33 CAD
CAAAA15958-06 CAAAA15958-14 CAAAA15958-22
Thiomorpholine 98%
Thiomorpholine

MDL: MFCD00005974 Beilstein: 102550 Miscible with water


Purity: 98% CAS No.: 123-90-0 Molecular Formula: C4H9NS Formula Weight: 103.19 Flash Point: 67°(153°F) Refractive Index: 1.5384 Boiling Point: 175-177° Solubility: Miscible with water Beilstein Reference: 102550 Merck Reference: 14,9300 UN No.: 3267 MDL No.: MFCD00005974

Formula: C₄H₉NS
Boiling Pt: 175…177 °C
Density: 1.086
Flash Pt: 67 °C(153 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005974
CAS Number: 123-90-0
EINECS: 204-660-2
UN: 3267
ADR: "8",III
Merck Index: 14,9300

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR