1-Decanethiol 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15913-AC
A15913-AE
A15913-AP
CAAAAA15913-ACEA
58.46
CAD
CAAAAA15913-AC
CAAAA15913-AE
CAAAA15913-AP
1-Decanethiol 96%
1-Decanethiol
Purity: 96%
CAS No.: 143-10-2
Molecular Formula: C10H22S
Formula Weight: 174.35
Flash Point: 98°(208°F)
Refractive Index: 1.4565
Odor: Unpleasant
Melting Point: -26°
Boiling Point: 237°
Beilstein Reference: 1735223
MDL No.: MFCD00004884
Formula:
C₁₀H₂₂S Boiling Pt: 237 °C Melting Pt: –26 °C Density: 0.843 Flash Pt: 98 °C(208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004884 CAS Number: 143-10-2 EINECS: 205-584-2 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...