1-Iodo-2-methylpropane 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with copper
A15883-14
A15883-22
CAAAAA15883-14EA
151.68
CAD
CAAAAA15883-14
CAAAAA15883-22
1-Iodo-2-methylpropane 97% stabilized
1-Iodo-2-methylpropane
Purity: 98%
CAS No.: 513-38-2
Molecular Formula: C4H9I
Formula Weight: 184.02
Flash Point: 12°(53°F)
Refractive Index: 1.4960
Melting Point: -93°
Boiling Point: 120-121°
Beilstein Reference: 1730927
Merck Reference: 14,5144
UN No.: 2391
MDL No.: MFCD00001084
Formula:
C₄H₉I Boiling Pt: 120…121 °C Melting Pt: –93 °C Density: 1.599 Flash Pt: 12 °C (53 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001084 CAS Number: 513-38-2 EINECS: 208-160-5 UN: 2391 ADR: 3,II Merck Index: 14,5144 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...