Cadmium(II) carbonate 97%

Supplier: Thermo Scientific Chemicals
Danger

A15812-0B A15812-18 A15812-30
CAAAAA15812-0BEA 377.62 CAD
CAAAAA15812-0B CAAAAA15812-18 CAAAAA15812-30
Cadmium(II) carbonate 97%
Cadmium (II) carbonate

Purity: 97% CAS No.: 513-78-0 Molecular Formula: CdCO3 Formula Weight: 172.42 Odor: Odorless Melting Point: 500° dec. UN No.: 2570 MDL No.: MFCD00010918

Formula: CCdO₃
MW: 172.42 g/mol
Melting Pt: 500 °C (dec.)
Density: 4.26
Storage Temperature: Ambient
MDL Number: MFCD00010918
CAS Number: 513-78-0
EINECS: 208-168-9
UN: 2570
ADR: 6.1,III
Merck Index: 14,1616

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR