Nitrosium tetrafluoroborate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nitrosyl tetrafluoroborate, Nitrosonium tetrafluoroborate

A15806-04 A15806-09 A15806-18
CAAAAA15806-04EA 75.05 CAD
CAAAAA15806-04 CAAAAA15806-09 CAAAAA15806-18
Nitrosium tetrafluoroborate 98%
Nitrosium tetrafluoroborate

Purity: 98% CAS No.: 14635-75-7 Molecular Formula: NOBF4 Formula Weight: 116.82 Storage Temperature: 2°C to 8°C Odor: Irritating Boiling Point: 250°/0.01mm subl. Merck Reference: 14,6649 UN No.: 3260 MDL No.: MFCD00011433

Formula: BF₄NO
MW: 116.81 g/mol
Boiling Pt: 250 °C/0.01 mm (subl.)
Melting Pt: 250 °C (subl.)
Density: 2.185
Storage Temperature: Refrigerator
MDL Number: MFCD00011433
CAS Number: 14635-75-7
EINECS: 238-679-2
UN: 3260
ADR: 8,III
Merck Index: 14,6649

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR