4-Methoxybenzaldehyde dimethyl acetal 98%

Supplier: Thermo Scientific Chemicals

Synonyms: p-(Dimethoxymethyl)anisole, P-anisaldehyde dimethyl acetal, p-Anisaldehyde dimethyl acetal, 1-(Dimethoxymethyl)-4-methoxybenzene, 4-Methoxy-α,α-dimethoxytoluene

A15793-14 A15793-36 A15793-30
CAAAA15793-14EA 93.06 CAD
CAAAA15793-14 CAAAA15793-36 AAA15793-30
4-Methoxybenzaldehyde dimethyl acetal 98%
4-Methoxybenzaldehyde dimethyl acetal

Purity: 98% CAS No.: 2186-92-7 Molecular Formula: C10H14O3 Formula Weight: 182.22 Flash Point: 97°(206°F) Refractive Index: 1.5055 Odor: Pleasant Boiling Point: 253° Beilstein Reference: 2048930 MDL No.: MFCD00036507

Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
Boiling Pt: 253 °C
Density: 1.068
Flash Pt: 97 °C(206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036507
CAS Number: 2186-92-7
EINECS: 218-577-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR