Diphenyl ether 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diphenyl oxide, Phenyl ether, Phenoxybenzene
A15791-0E
A15791-30
A15791-36
CAAAAA15791-0EEA
175.38
CAD
CAAAAA15791-0E
CAAAAA15791-30
CAAAAA15791-36
Diphenyl ether 99%
Diphenyl ether
Purity: 99%
CAS No.: 101-84-8
Molecular Formula: C12H10O
Formula Weight: 170.21
Flash Point: 125°(257°F)
Refractive Index: 1.5805
Odor: Aromatic
Melting Point: 26-29°
Boiling Point: 258-260°
Beilstein Reference: 1364620
Merck Reference: 14,7288
UN No.: 3077
MDL No.: MFCD00003034
Formula:
C₁₂H₁₀O MW: 170.21 g/mol Boiling Pt: 498 Melting Pt: 26…29 °C Density: 1.071 Flash Pt: 125 °C (257 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003034 CAS Number: 101-84-8 EINECS: 202-981-2 UN: 3077 ADR: 9 III Merck Index: 14,7288 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...