2,2'-Bipyridyl 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15782-14
A15782-22
A15782-36
CAAAAA15782-14EA
132.25
CAD
CAAAAA15782-14
CAAAAA15782-22
CAAAAA15782-36
2,2'-Bipyridyl 99+%
2,2'-Bipyridyl
Reagent for iron determinations
Purity: 99+%
CAS No.: 366-18-7
Molecular Formula: C10H8N2
Formula Weight: 156.19
Flash Point: 121°(249°F)
Melting Point: 70°
Boiling Point: 272-273°
Beilstein Reference: 113089
Merck Reference: 14,3347
UN No.: 2811
MDL No.: MFCD00006212
Formula:
C₁₀H₈N₂ Boiling Pt: 272…273 °C Melting Pt: 70 °C Density: 1.326 g/cm³ (-150 °C) Flash Pt: 121 °C (249 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006212 CAS Number: 366-18-7 EINECS: 206-674-4 UN: 2811 ADR: 6.1,III Merck Index: 14,3347 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...