α-Chloro-4-nitrotoluene 99%

Supplier: Thermo Scientific Chemicals
Danger

A15749-09 A15749-18 A15749-30
CAAAA15749-09EA 48.98 CAD
CAAAA15749-09 CAAAA15749-18 CAAAA15749-30
α-Chloro-4-nitrotoluene 99%
α-Chloro-4-nitrotoluene

Purity: 99% CAS No.: 100-14-1 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: 70-74° Beilstein Reference: 387187 UN No.: 3261 MDL No.: MFCD00007374

Formula: C₇H₆ClNO₂
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 70…74 °C
Density: 1.5647 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007374
CAS Number: 100-14-1
EINECS: 202-822-7
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR