1-Bromo-4-chloro-3-fluorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-1-chloro-2-fluorobenzene

A15745-14 A15745-22
CAAAAA15745-06EA 54.59 CAD
CAAAAA15745-06 CAAAAA15745-14 CAAAAA15745-22
1-Bromo-4-chloro-3-fluorobenzene 98%
1-Bromo-4-chloro-3-fluorobenzene

Purity: 98% CAS No.: 60811-18-9 Molecular Formula: C6H3BrClF Formula Weight: 209.45 Refractive Index: 1.5550 Boiling Point: 62-64°/9mm MDL No.: MFCD00042519

Formula: C₆H₃BrClF
MW: 209.44 g/mol
Boiling Pt: 62…64 °C (9 mmHg)
Density: 1.710
Storage Temperature: Ambient
MDL Number: MFCD00042519
CAS Number: 60811-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR