Ethylene carbonate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15735-0C
A15735-0E
A15735-36
CAAAAA15735-0CEA
589.34
CAD
CAAAAA15735-0C
CAAAAA15735-0E
CAAAAA15735-36
Ethylene carbonate 99%
Ethylene carbonate
Solvent for many polymers and resins, plasticisers, rubbers and textiles.
Purity: 99%
CAS No.: 96-49-1
Molecular Formula: C3H4O3
Formula Weight: 88.06
Flash Point: 160°(320°F)
Melting Point: 35-37°
Boiling Point: 244-245°
Solubility: Miscible (40%) with water, alchohol ethyl acetate, benzene and chloroform. Soluble in ether, n-Butanol, and carbon tetrachloride.
Beilstein Reference: 106249
MDL No.: MFCD00005382
Formula:
C₃H₄O₃ Boiling Pt: 244…245 °C Melting Pt: 35…37 °C Density: 1.320 Flash Pt: 160 °C(320 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005382 CAS Number: 96-49-1 EINECS: 202-510-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...