4-(Benzyloxy)benzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A15715-03 A15715-06 A15715-14
CAAAA15715-03EA 37.13 CAD
CAAAA15715-03 CAAAA15715-06 CAAAA15715-14
4-(Benzyloxy)benzoic acid 98%
4-(Benzyloxy)benzoic acid

Purity: 98% CAS No.: 1486-51-7 Molecular Formula: C14H12O3 Formula Weight: 228.25 Melting Point: 187-191° Beilstein Reference: 2111640

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Melting Pt: 187…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00016527
CAS Number: 1486-51-7
EINECS: 216-066-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR