Trisodium phosphate dodecahydrate 97%

Supplier: Thermo Scientific Chemicals
Warning

A15713-0C A15713-0E A15713-36
CAAAAA15713-0CEA 1131.28 CAD
CAAAAA15713-0C CAAAAA15713-0E CAAAAA15713-36
Trisodium phosphate dodecahydrate 97%
tri-Sodium phosphate dodecahydrate

Purity: 97% CAS No.: 10101-89-0 Molecular Formula: Na3O4P.12H2O Formula Weight: 380.12 (163.94anhy) Odor: Odorless pH: 12 Melting Point: ca 75° dec. Merck Reference: 14,8662 UN No.: 3262 MDL No.: MFCD00149198

Formula: Na₃PO₄·12H₂O
Melting Pt: ∼ 75 °C (dec.)
Density: 1.62
Storage Temperature: Ambient
MDL Number: MFCD00149198
CAS Number: 10101-89-0
EINECS: 231-509-8
ADR: 8,III
REACH: 01-2119489800-32-xxxx
Merck Index: 14,8662

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR