Sodium tellurite(IV) 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium tellurite, Disodium trioxotellurate

A15633-18 A15633-30
CAAAAA15633-0BEA 4571.48 CAD
CAAAAA15633-0B CAAAAA15633-18 CAAAAA15633-30
Sodium tellurite(IV) 97%
Sodium tellurite(IV)

Purity: 97% CAS No.: 10102-20-2 Molecular Formula: Na2O3Te Formula Weight: 221.58 Merck Reference: 14,8685 UN No.: 3284 MDL No.: MFCD00036141

Formula: Na₂O₃Te
Melting Pt: 710 °C
Storage Temperature: Ambient
MDL Number: MFCD00036141
CAS Number: 10102-20-2
EINECS: 233-268-4
UN: 3284
ADR: 6.1,III
Merck Index: 14,8685

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR