1-Naphthaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: α-Naphthaldehyde

A15610-0B A15610-18 A15610-30
CAAAAA15610-0BEA 782.1 CAD
CAAAAA15610-0B CAAAAA15610-18 CAAAAA15610-30
1-Naphthaldehyde 97%
1-Naphthaldehyde

Purity: 97% CAS No.: 66-77-3 Molecular Formula: C11H8O Formula Weight: 156.18 Flash Point: 112°(233°F) Refractive Index: 1.6530 Odor: Pungent Melting Point: 1-2° Boiling Point: 160-161°/15mm Beilstein Reference: 386082 MDL No.: MFCD00004003

Formula: C₁₁H₈O
MW: 156.18 g/mol
Boiling Pt: 160…161 °C/15 mm
Melting Pt: 1…2 °C
Density: 1.148
Flash Pt: 112 °C(233 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004003
CAS Number: 66-77-3
EINECS: 200-633-4
UN: 3334

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR