Potassium tetrafluoroborate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15605-0B
A15605-0I
A15605-30
CAAAAA15605-0BEA
153.58
CAD
CAAAAA15605-0B
CAAAAA15605-0I
CAAAAA15605-30
Potassium tetrafluoroborate 98%
Potassium tetrafluoroborate
Purity: 98%
CAS No.: 14075-53-7
Molecular Formula: BF4K
Formula Weight: 125.91
Refractive Index: 1.3245
Melting Point: 529°
Boiling Point: dec.
Merck Reference: 14,7686
MDL No.: MFCD00011395
Formula:
BF₄K Boiling Pt: Dec. Melting Pt: 529 °C Density: 2.50 Storage Temperature: Ambient |
MDL Number:
MFCD00011395 CAS Number: 14075-53-7 EINECS: 237-928-2 Merck Index: 14,7686 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...